2-(4-Bromo-2-fluorophenoxy)-4-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC300643-1G PC300643-10G PC300643-5G
APOSPC300643-1GEA 288 EUR
APOSPC300643-1G APOSPC300643-10G APOSPC300643-5G
2-(4-Bromo-2-fluorophenoxy)-4-(trifluoromethyl)pyrimidine
2-(4-Bromo-2-fluorophenoxy)-4-(trifluoromethyl)pyrimidine
Formel: C₁₁H₅BrF₄N₂O
Molekulargewicht: 337.068 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11520136
CAS-Nummer: 1227955-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR