8-Methoxy-4-(trifluoromethyl)quinolin-2(1H)-one

Lieferant: Apollo Scientific

PC300630-1G PC300630-250MG
APOSPC300630-1GEA 238 EUR
APOSPC300630-1G APOSPC300630-250MG
8-Methoxy-4-(trifluoromethyl)quinolin-2(1H)-one
8-Methoxy-4-(trifluoromethyl)quinolin-2(1H)-one
Formel: C₁₁H₈F₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16631270
CAS-Nummer: 503147-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR