1,1,1,3,3,3-Hexafluoro-2-methylpropane 97%
Lieferant: Apollo Scientific
PC2956-25G
PC2956-5G
PC2956-100G
APOSPC2956-25GEA
765
EUR
APOSPC2956-25G
APOSPC2956-5G
APOSPC2956-100G
1,1,1,3,3,3-Hexafluoro-2-methylpropane 97%
1,1,1,3,3,3-Hexafluoro-2-methylpropane
Formel:
C₄H₄F₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03094211 CAS-Nummer: 382-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...