2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide
Lieferant: Apollo Scientific
|
Achtung
|
PC28280-1G
PC28280-250MG
APOSPC28280-1GEA
98.6
EUR
APOSPC28280-1G
APOSPC28280-250MG
2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide
2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide
Formel:
C₁₀H₁₀ClFN₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31812107 CAS-Nummer: 2379917-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...