8-Bromo-6-(trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
Lieferant: Apollo Scientific
PC28251-1G
PC28251-250MG
APOSPC28251-1GEA
522
EUR
APOSPC28251-1G
APOSPC28251-250MG
8-Bromo-6-(trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
8-Bromo-6-(trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
Formel:
C₁₁H₄BrF₃O₅ |
MDL Number:
MFCD31793557 CAS-Nummer: 2379918-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...