8-Fluoro-6-(trifluoromethyl)quinolin-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
PC28242-1G
PC28242-250MG
APOSPC28242-1GEA
241
EUR
APOSPC28242-1G
APOSPC28242-250MG
8-Fluoro-6-(trifluoromethyl)quinolin-4-ol
8-Fluoro-6-(trifluoromethyl)quinolin-4-ol
Formel:
C₁₀H₅F₄NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28010364 CAS-Nummer: 1065092-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...