3-Oxo-N-[2-(trifluoromethyl)phenyl]butanamide 95%
Lieferant: Apollo Scientific
PC27951-1G
PC27951-5G
APOSPC27951-1GEA
339
EUR
APOSPC27951-1G
APOSPC27951-5G
3-Oxo-N-[2-(trifluoromethyl)phenyl]butanamide 95%
3-Oxo-N-[2-(trifluoromethyl)phenyl]butanamide
Formel:
C₁₁H₁₀F₃NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00231302 CAS-Nummer: 81761-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...