3-Bromo-5-nitrobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27896-1G PC27896-250MG
APOSPC27896-1GEA 609 EUR
APOSPC27896-1G APOSPC27896-250MG
3-Bromo-5-nitrobenzyl fluoride 95%
3-Bromo-5-nitrobenzyl fluoride
Formel: C₇H₅BrFNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31664893
CAS-Nummer: 2244084-96-4
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR