Fmoc-α-Methyl-L-4-fluorophenylalanine

Lieferant: Apollo Scientific

PC27888-1G PC27888-250MG
APOSPC27888-1GEA 546 EUR
APOSPC27888-1G APOSPC27888-250MG
Fmoc-α-Methyl-L-4-fluorophenylalanine
Fmoc-α-Methyl-L-4-fluorophenylalanine
Formel: C₂₅H₂₂FNO₄
Molekulargewicht: 419.445 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08061624
CAS-Nummer: 1175838-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR