3-Bromo-5-fluoro-2-hydroxybenzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC2607-1G
PC2607-250MG
APOSPC2607-1GEA
52.2
EUR
APOSPC2607-1G
APOSPC2607-250MG
3-Bromo-5-fluoro-2-hydroxybenzaldehyde
3-Bromo-5-fluoro-2-hydroxybenzaldehyde
Formel:
C₇H₄BrFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08458073 CAS-Nummer: 178546-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...