α,N-Dimethyl-3-fluorobenzylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Fluorophenyl)-N-methylethylamine
PC2543-1G
PC2543-5G
APOSPC2543-1GEA
94.4
EUR
APOSPC2543-1G
APOSPC2543-5G
α,N-Dimethyl-3-fluorobenzylamine
α,N-Dimethyl-3-fluorobenzylamine
Formel:
C₉H₁₂FN Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07784285 CAS-Nummer: 820209-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...