Potassium 4-(trifluoromethoxy) phenyltrifluoroborate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(4-(trifluoromethoxy)phenyl)borate
PC210104-1G
PC210104-5G
APOSPC210104-1GEA
132
EUR
APOSPC210104-1G
APOSPC210104-5G
Potassium 4-(trifluoromethoxy) phenyltrifluoroborate 95%
Potassium 4-(trifluoromethoxy) phenyltrifluoroborate
Formel:
C₇H₄BF₆O·K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18861588 CAS-Nummer: 1394827-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...