8-Fluoroquinolin-2-amine 95+%

Lieferant: Apollo Scientific

PC201122-100MG PC201122-250MG
APOSPC201122-100MGEA 300 EUR
APOSPC201122-100MG APOSPC201122-250MG
8-Fluoroquinolin-2-amine 95+%
8-Fluoroquinolin-2-amine
Formel: C₉H₇FN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19678631
CAS-Nummer: 1342095-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR