Methyl 3-amino-4-(trifluoromethyl)benzoate 98%

Lieferant: Apollo Scientific

PC201106-1G PC201106-250MG PC201106-5G
APOSPC201106-1GEA 321 EUR
APOSPC201106-1G APOSPC201106-250MG APOSPC201106-5G
Methyl 3-amino-4-(trifluoromethyl)benzoate 98%
Methyl 3-amino-4-(trifluoromethyl)benzoate
Formel: C₉H₈F₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14697987
CAS-Nummer: 126541-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR