5-Nitro-2-(trifluoromethyl)phenol 95%

Lieferant: Apollo Scientific
Achtung

PC200419-1G PC200419-250MG PC200419-500MG
APOSPC200419-1GEA 609 EUR
APOSPC200419-1G APOSPC200419-250MG APOSPC200419-500MG
5-Nitro-2-(trifluoromethyl)phenol 95%
5-Nitro-2-(trifluoromethyl)phenol
Formel: C₇H₄F₃NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16606321
CAS-Nummer: 612498-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR