Potassium (3-aminophenyl)trifluoroboranuide
Lieferant: Apollo Scientific
Synonyme:
Potassium (3-aminophenyl)trifluoroborate
PC200388-1G
PC200388-5G
APOSPC200388-1GEA
97.8
EUR
APOSPC200388-1G
APOSPC200388-5G
Potassium (3-aminophenyl)trifluoroboranuide
Potassium (3-aminophenyl)trifluoroboranuide
Formel:
C₆H₆BF₃N·K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09992971 CAS-Nummer: 1160186-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...