1,4-Dipentafluorophenyl benzene-1,4-dicarboxylate
Lieferant: Apollo Scientific
PC200026-1G
PC200026-500MG
APOSPC200026-1GEA
118
EUR
APOSPC200026-1G
APOSPC200026-500MG
1,4-Dipentafluorophenyl benzene-1,4-dicarboxylate
1,4-Dipentafluorophenyl benzene-1,4-dicarboxylate
Formel:
C₂₀H₄F₁₀O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20274437 CAS-Nummer: 133921-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...