3,5-Difluoro-D-phenylalanine, N-FMOC protected

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Phe(3,5-DiF)-OH, (2R)-3-(3,5-Difluorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,5-difluorophenyl)propanoic acid

PC1517-1G PC1517-250MG
APOSPC1517-1GEA 92.8 EUR
APOSPC1517-1G APOSPC1517-250MG
3,5-Difluoro-D-phenylalanine, N-FMOC protected
3,5-Difluoro-D-phenylalanine, N-FMOC protected
Formel: C₂₄H₁₉F₂NO₄
Molekulargewicht: 423.409 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00797580
CAS-Nummer: 205526-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR