(Ir[dF(CF3)ppy]2(dtbpy))PF6 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine-N1,N1′]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-N]phenyl-C]Iridium(III) hexafluorophosphate
PC13675-100MG
PC13675-250MG
APOSPC13675-100MGEA
40.6
EUR
APOSPC13675-100MG
APOSPC13675-250MG
(Ir[dF(CF3)ppy]2(dtbpy))PF6 98%
(Ir[dF(CF3)ppy]2(dtbpy))PF6
Formel:
C₄₂H₃₆F₁₀IrN₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD27952556 CAS-Nummer: 870987-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...