4-Bromo-2,5-difluorotoluene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2,5-difluoro-4-methylbenzene

PC11267-1G PC11267-5G PC11267-250MG
APOSPC11267-1GEA 36.4 EUR
APOSPC11267-1G APOSPC11267-5G APOSPC11267-250MG
4-Bromo-2,5-difluorotoluene
4-Bromo-2,5-difluorotoluene
Formel: C₇H₅BrF₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07368736
CAS-Nummer: 252004-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR