4-Chloro-8-fluoro-7-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific
Achtung

PC110010-1G PC110010-250MG
APOSPC110010-1GEA 281 EUR
APOSPC110010-1G APOSPC110010-250MG
4-Chloro-8-fluoro-7-(trifluoromethyl)quinoline
4-Chloro-8-fluoro-7-(trifluoromethyl)quinoline
Formel: C₁₀H₄ClF₄N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32200322
CAS-Nummer: 693818-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR