3'-Bromo-2'-methoxy-2,2,2-trifluoroacetophenone 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-6-(trifluoroacetyl)anisole, 1-(3-Bromo-2-methoxyphenyl)-2,2,2-trifluoroethan-1-one
PC10679-1G
PC10679-250MG
PC10679-5G
APOSPC10679-1GEA
393
EUR
APOSPC10679-1G
APOSPC10679-250MG
APOSPC10679-5G
3'-Bromo-2'-methoxy-2,2,2-trifluoroacetophenone 97%
3'-Bromo-2'-methoxy-2,2,2-trifluoroacetophenone
Formel:
C₉H₆BrF₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26407434 CAS-Nummer: 1820639-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...