3-Bromo-2-fluoro-5-methylaniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC10464-1G
PC10464-250MG
APOSPC10464-1GEA
654
EUR
APOSPC10464-1G
APOSPC10464-250MG
3-Bromo-2-fluoro-5-methylaniline 95%
3-Bromo-2-fluoro-5-methylaniline
Formel:
C₇H₇BrFN Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22627835 CAS-Nummer: 1207319-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...