2,3,4,5,6-Pentafluorochalcone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Pentafluorophenyl)-1-phenylprop-2-en-1-one, 3-(Perfluorophenyl)-1-phenylprop-2-en-1-one
PC0826-1G
PC0826-5G
APOSPC0826-1GEA
69.6
EUR
APOSPC0826-1G
APOSPC0826-5G
2,3,4,5,6-Pentafluorochalcone
2,3,4,5,6-Pentafluorochalcone
Formel:
C₁₅H₇F₅O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00483321 CAS-Nummer: 15269-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...