2-Methyl-5-(trifluoromethyl)benzoyl chloride

Lieferant: Apollo Scientific
Gefahr

PC0466-1G PC0466-2.5G PC0466-5G
APOSPC0466-1GEA 34.8 EUR
APOSPC0466-1G APOSPC0466-2.5G APOSPC0466-5G
2-Methyl-5-(trifluoromethyl)benzoyl chloride
2-Methyl-5-(trifluoromethyl)benzoyl chloride
Formel: C₉H₆ClF₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631599
CAS-Nummer: 261952-08-3
UN-Nummer: 3264
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR