2-Amino-5-nitro-4-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC01523-1G
PC01523-250MG
PC01523-5G
APOSPC01523-1GEA
244
EUR
APOSPC01523-1G
APOSPC01523-250MG
APOSPC01523-5G
2-Amino-5-nitro-4-(trifluoromethyl)pyridine
2-Amino-5-nitro-4-(trifluoromethyl)pyridine
Formel:
C₆H₄F₃N₃O₂ Schmelzpunkt: 138…140 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28155019 CAS-Nummer: 1000152-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...