Methyl (3,5-dimethyl-1H-pyrazol-4-yl)acetate 96%
Lieferant: Apollo Scientific
OR965647-1G
OR965647-250MG
APOSOR965647-1GEA
632
EUR
APOSOR965647-1G
APOSOR965647-250MG
Methyl (3,5-dimethyl-1H-pyrazol-4-yl)acetate 96%
Methyl (3,5-dimethyl-1H-pyrazol-4-yl)acetate
Formel:
C₈H₁₂N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07685912 CAS-Nummer: 56699-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...