2-Methyl-1H-imidazole-1-acetic acid ethyl ester 95%
Lieferant: Apollo Scientific
OR965529-1G
OR965529-250MG
APOSOR965529-1GEA
328
EUR
APOSOR965529-1G
APOSOR965529-250MG
2-Methyl-1H-imidazole-1-acetic acid ethyl ester 95%
2-Methyl-1H-imidazole-1-acetic acid ethyl ester
Formel:
C₈H₁₂N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03248721 CAS-Nummer: 239065-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...