1,3-Diazaspiro[4.4]nonane-2,4-dione 95%
Lieferant: Apollo Scientific
OR965025-1G
OR965025-5G
APOSOR965025-1GEA
46.4
EUR
APOSOR965025-1G
APOSOR965025-5G
1,3-Diazaspiro[4.4]nonane-2,4-dione 95%
1,3-Diazaspiro[4.4]nonane-2,4-dione
Formel:
C₇H₁₀N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00022403 CAS-Nummer: 699-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...