4-Amino-2-(methylthio)pyridine 95%
Lieferant: Apollo Scientific
OR964583-1G
OR964583-250MG
APOSOR964583-1GEA
403
EUR
APOSOR964583-1G
APOSOR964583-250MG
4-Amino-2-(methylthio)pyridine 95%
4-Amino-2-(methylthio)pyridine
Formel:
C₆H₈N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16084526 CAS-Nummer: 59243-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...