(3-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%

Lieferant: Apollo Scientific

OR962884-1G OR962884-250MG
APOSOR962884-1GEA 290 EUR
APOSOR962884-1G APOSOR962884-250MG
(3-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%
(3-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol
Formel: C₁₁H₁₂N₂O
Molekulargewicht: 188.226 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11870095
CAS-Nummer: 167758-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR