2-(2-Methoxyethoxy)-benzonitrile 97%

Lieferant: Apollo Scientific

OR962770-1G OR962770-5G
APOSOR962770-1GEA 248 EUR
APOSOR962770-1G APOSOR962770-5G
2-(2-Methoxyethoxy)-benzonitrile 97%
2-(2-Methoxyethoxy)-benzonitrile
Formel: C₁₀H₁₁NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09907639
CAS-Nummer: 540753-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR