DL-3-(BOC-Amino)-1-butanol 97%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (4-hydroxybutan-2-yl)carbamate
OR962733-1G
OR962733-5G
APOSOR962733-1GEA
59.6
EUR
APOSOR962733-1G
APOSOR962733-5G
DL-3-(BOC-Amino)-1-butanol 97%
DL-3-(BOC-Amino)-1-butanol
Formel:
C₉H₁₉NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09031504 CAS-Nummer: 146514-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...