2-[(4-Chlorophenyl)amino]acetic acid 98%
Lieferant: Apollo Scientific
Synonyme:
N-(4-Cl-Phenyl)-Gly-OH, 2-((4-Chlorophenyl)amino)acetic acid
OR962513-1G
OR962513-5G
APOSOR962513-1GEA
203
EUR
APOSOR962513-1G
APOSOR962513-5G
2-[(4-Chlorophenyl)amino]acetic acid 98%
2-[(4-Chlorophenyl)amino]acetic acid
Formel:
C₈H₈ClNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00462901 CAS-Nummer: 5465-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...