N-Boc-2-amino-2-phenylethylamine 97%

Lieferant: Apollo Scientific

OR961674-1G OR961674-500MG
APOSOR961674-1GEA 731 EUR
APOSOR961674-1G APOSOR961674-500MG
N-Boc-2-amino-2-phenylethylamine 97%
N-Boc-2-amino-2-phenylethylamine
Formel: C₁₃H₂₀N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06245566
CAS-Nummer: 174885-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR