4-Phenyloxan-4-ol 95%
Lieferant: Apollo Scientific
OR960325-1G
OR960325-250MG
APOSOR960325-1GEA
435
EUR
APOSOR960325-1G
APOSOR960325-250MG
4-Phenyloxan-4-ol 95%
4-Phenyloxan-4-ol
Formel:
C₁₁H₁₄O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00729123 CAS-Nummer: 81462-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...