tert-Butyl N,N-diethylcarbamate 98%
Lieferant: Apollo Scientific
OR960322-1G
OR960322-5G
APOSOR960322-1GEA
219
EUR
APOSOR960322-1G
APOSOR960322-5G
tert-Butyl N,N-diethylcarbamate 98%
tert-Butyl N,N-diethylcarbamate
Formel:
C₉H₁₉NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20929510 CAS-Nummer: 51170-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...