5-Bromo-4-chloro-2-methoxybenzonitrile 98%

Lieferant: Apollo Scientific

OR960009-1G OR960009-5G
APOSOR960009-1GEA 219 EUR
APOSOR960009-1G APOSOR960009-5G
5-Bromo-4-chloro-2-methoxybenzonitrile 98%
5-Bromo-4-chloro-2-methoxybenzonitrile
Formel: C₈H₅BrClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16873032
CAS-Nummer: 1239838-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR