6-Chloro-2-(dimethylamino)nicotinaldehyde 98%

Lieferant: Apollo Scientific

OR959963-1G OR959963-5G
APOSOR959963-1GEA 290 EUR
APOSOR959963-1G APOSOR959963-5G
6-Chloro-2-(dimethylamino)nicotinaldehyde 98%
6-Chloro-2-(dimethylamino)nicotinaldehyde
Formel: C₈H₉ClN₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21333091
CAS-Nummer: 1233698-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR