Methyl 5-amino-2-chloropyridine-3-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-amino-2-chloronicotinate

OR959677-1G OR959677-250MG
APOSOR959677-1GEA 273 EUR
APOSOR959677-1G APOSOR959677-250MG
Methyl 5-amino-2-chloropyridine-3-carboxylate 98%
Methyl 5-amino-2-chloropyridine-3-carboxylate
Formel: C₇H₇ClN₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11848216
CAS-Nummer: 886435-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR