2-Bromo-5-chloro-4-methylpyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR959457-1G
OR959457-25G
OR959457-5G
APOSOR959457-1GEA
86.1
EUR
APOSOR959457-1G
APOSOR959457-25G
APOSOR959457-5G
2-Bromo-5-chloro-4-methylpyridine 98%
2-Bromo-5-chloro-4-methylpyridine
Formel:
C₆H₅BrClN Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD11846275 CAS-Nummer: 885267-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...