3-T-Butyl-5-hydroxybenzoic acid 98%

Lieferant: Apollo Scientific

OR957791-1G OR957791-5G
APOSOR957791-1GEA 449 EUR
APOSOR957791-1G APOSOR957791-5G
3-T-Butyl-5-hydroxybenzoic acid 98%
3-T-Butyl-5-hydroxybenzoic acid
Formel: C₁₁H₁₄O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09749938
CAS-Nummer: 49843-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR