2-Methyl-4-phenylpyridine 98%
Lieferant: Apollo Scientific
OR955415-1G
OR955415-250MG
APOSOR955415-1GEA
574
EUR
APOSOR955415-1G
APOSOR955415-250MG
2-Methyl-4-phenylpyridine 98%
2-Methyl-4-phenylpyridine
Formel:
C₁₂H₁₁N Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04114154 CAS-Nummer: 15032-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...