2-Hydroxy-3-phenylpyridine 95%
Lieferant: Apollo Scientific
Synonyme:
3-Phenylpyridin-2-ol
OR954647-1G
OR954647-250MG
APOSOR954647-1GEA
444
EUR
APOSOR954647-1G
APOSOR954647-250MG
2-Hydroxy-3-phenylpyridine 95%
2-Hydroxy-3-phenylpyridine
Formel:
C₁₁H₉NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04114134 CAS-Nummer: 24228-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...