5-Cyclohexyl-1,3,4-oxadiazole-2-thiol 95%

Lieferant: Apollo Scientific

OR953654-1G OR953654-5G
APOSOR953654-1GEA 290 EUR
APOSOR953654-1G APOSOR953654-5G
5-Cyclohexyl-1,3,4-oxadiazole-2-thiol 95%
5-Cyclohexyl-1,3,4-oxadiazole-2-thiol
Formel: C₈H₁₂N₂OS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06655106
CAS-Nummer: 173589-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR