4-Chlorobutane-1-sulphonyl chloride 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chloro-1-butylsulphonyl Chloride

OR953416-1G OR953416-5G
APOSOR953416-1GEA 63.8 EUR
APOSOR953416-1G APOSOR953416-5G
4-Chlorobutane-1-sulphonyl chloride 96%
4-Chlorobutane-1-sulphonyl chloride
Formel: C₄H₈Cl₂O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09952390
CAS-Nummer: 1633-84-7
UN-Nummer: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR