cis-Ethyl 2-hydroxy-cyclohexanecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
cis-Ethyl 2-hydroxycyclohexanecarboxylate
OR953021-1G
OR953021-250MG
APOSOR953021-1GEA
273
EUR
APOSOR953021-1G
APOSOR953021-250MG
cis-Ethyl 2-hydroxy-cyclohexanecarboxylate 95%
cis-Ethyl 2-hydroxy-cyclohexanecarboxylate
Formel:
C₉H₁₆O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00144282 CAS-Nummer: 6149-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...