Methyl 2-hexenoate 98%
Lieferant: Apollo Scientific
OR952905-100G
OR952905-25G
APOSOR952905-100GEA
127
EUR
APOSOR952905-100G
APOSOR952905-25G
Methyl 2-hexenoate 98%
Methyl 2-hexenoate
Formel:
C₇H₁₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00048798 CAS-Nummer: 2396-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...