1-(tert-Butyl)-3-methoxybenzene 95%

Lieferant: Apollo Scientific

OR952242-1G OR952242-250MG OR952242-5G
APOSOR952242-1GEA 89.9 EUR
APOSOR952242-1G APOSOR952242-250MG APOSOR952242-5G
1-(tert-Butyl)-3-methoxybenzene 95%
1-(tert-Butyl)-3-methoxybenzene
Formel: C₁₁H₁₆O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD15527519
CAS-Nummer: 33733-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR