Methyl 3-methylisoxazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR950812-1G OR950812-250MG
APOSOR950812-1GEA 560 EUR
APOSOR950812-1G APOSOR950812-250MG
Methyl 3-methylisoxazole-5-carboxylate 95%
Methyl 3-methylisoxazole-5-carboxylate
Formel: C₆H₇NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10699057
CAS-Nummer: 1004-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR